Kits: the case of the invisible bands, Ready-to-Load™
Supplier: EDVOTEK
S-52
763-0009EA 0 EUR
763-0009
Kits: the case of the invisible bands, Ready-to-Load™
Educational Materials Biology Educational Materials Genetics Learning Activities
Experiment using fluorescence and electrophoresis to solve the mystery of the invisible bands.
  • For 10 lab groups
  • Complete in 45 minutes
Also required: Electrophoresis tank, power supply and long wave UV lamp.
Delivery information: Kit includes instructions, Ready-to-Load™ dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, and microtipped transfer pipettes.