Chlorosulphuric acid for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Chlorosulphonic acid

M-Clarity™ quality level = MQ200

8.00220.9050
8.00220.9050EA 0 EUR
8.00220.9050
Chlorosulphuric acid for synthesis, Sigma-Aldrich®
Chlorosulphuric acid
Formula: HClO₃S
MW: 116.52 g/mol
Boiling Pt: 152
Melting Pt: –80 °C
Density: 1.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011523
CAS Number: 7790-94-5
EINECS: 232-234-6
UN: 1754
ADR: 8 I
Merck Index: 12,02218

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR