Glutaric acid for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 1,5-Pentanedioic acid

M-Clarity™ quality level = MQ200

8.00295.0100 8.00295.0250 8.00295.9025 8.00295.1000
8.00295.0100EA 34.1 EUR
8.00295.0100 8.00295.0250 8.00295.9025 8.00295.1000
Glutaric acid for synthesis, Sigma-Aldrich®
Glutaric acid
Formula: C₅H₈O₄
MW: 132.12 g/mol
Boiling Pt: 302
Melting Pt: 98
Density: 1.43 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004410
CAS Number: 110-94-1
EINECS: 203-817-2
UN: 3261
ADR: 8 II
Merck Index: 13,04486

Order Now

Specification Test Results

Solubility 640 g/l (20 °C)
Melting Point 98 °C
Molar Mass 132.11 g/mol
Boiling Point 302 - 304 °C (1013 hPa) (slow decomposition)
Vapor Pressure 0.022 hPa (18.5 °C)
Density 1.43 g/cm3 (15 °C)
Synonyms Pentanedioic acid
Hill Formula C5H8O4
Chemical Formula HOOC(CH2)3COOH


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR