L(-)-Malic acid for racemat separation, for synthesis, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: L(-)-Malic acid, (S)-(-)-Hydroxy-1,4-butanedioic acid, (S)-(-)-Hydroxysuccinic acid, (S)-2-hydroxysuccinic acid, L(-)-Hydroxysuccinic acid, L(-)-Hydroxy-1,4-butanedioic acid, L-(–)-Malic Acid

M-Clarity™ quality level = MQ200

8.00384.0010 8.00384.0050 8.00384.1000 8.00384.0500 8.00384.0005 8.00384.0000
8.00384.0010EA 31 EUR
8.00384.0010 8.00384.0050 8.00384.1000 8.00384.0500 8.00384.0005 8.00384.0000
L(-)-Malic acid for racemat separation, for synthesis, Sigma-Aldrich®
L(-)-Malic acid
Formula: C₄H₆O₅
MW: 134.09 g/mol
Boiling Pt: 322
Melting Pt: 101…104 °C
Density: 1.595 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064213
CAS Number: 97-67-6
EINECS: 202-601-5
Merck Index: 13,05730

Order Now

Specification Test Results

Solubility 160 g/l (20 °C)
Melting Point 101 - 104 °C
Molar Mass 134.08 g/mol
Bulk Density 600 kg/m3
Boiling Point 140 °C (decomposition)
Density 1.60 g/cm3 (20 °C)
pH 2.2 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR