Dimethyl malonate ≥99.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

8.00899.0100 8.00899.0500 8.00899.1000 8.00899.9025
8.00899.0100EA 28.6 EUR
8.00899.0100 8.00899.0500 8.00899.1000 8.00899.9025
Dimethyl malonate ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
Dimethyl malonate
Formula: C₅H₈O₄
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: –62 °C
Density: 1,15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008460
CAS Number: 108-59-8
EINECS: 203-597-8
Merck Index: 12,06169

Order Now

Specification Test Results

Assay (GC, area%) ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C) 1.151 - 1.155
Identity (IR) passes test
Boiling Point 181 - 183 °C (1013 hPa)
Vapor Pressure 0.15 hPa (20 °C)
Flash Point 85 °C
Refractive Index 1.4135 (20 °C, 589 nm)
Explosion Limit 1.3 - 17.4 %(V)
Density 1.15 g/cm3 (20 °C)
Ignition Point 440 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR