Ethylenediammonium dichloride, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

8.00948.0005 8.00948.0250 8.00948.9050
8.00948.0005EA 26.8 EUR
8.00948.0005 8.00948.0250 8.00948.9050
Ethylenediammonium dichloride, Sigma-Aldrich®
Ethylenediammonium dichloride
Formula: C₂H₁₀Cl₂N₂
Storage Temperature: Ambient
MDL Number: MFCD00012524
CAS Number: 333-18-6
EINECS: 206-369-6

Order Now

Specification Test Results

Solubility 300 g/l (20 °C)
Melting Point 300 °C
Molar Mass 133.02 g/mol
Bulk Density 650 kg/m3
pH 5 (100 g/l, H2O, 20 °C)
Synonyms 1,2-Diaminoethane dihydrochloride, Ethylenediamine dihydrochloride
Hill Formula C2H10Cl2N2
Chemical Formula H2NCH2CH2NH2 * 2HCl


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR