Aluminium bromide for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Aluminum bromide

M-Clarity™ quality level = MQ200

8.01068.0100 8.01068.0025 8.01068.2500
8.01068.0100EA 291 EUR
8.01068.0100 8.01068.0025 8.01068.2500
Aluminium bromide for synthesis, Sigma-Aldrich®
Aluminium bromide
Formula: AlBr₃
MW: 266.69 g/mol
Boiling Pt: 265
Melting Pt: 97
Density: 3.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003421
CAS Number: 7727-15-3
EINECS: 231-779-7
UN: 1725
ADR: 8 II
Merck Index: 13,00330

Order Now

Specification Test Results

Solubility (20 °C) (rigorous decomposition)
Melting Point 97 °C
Molar Mass 266.69 g/mol
Boiling Point 250 - 270 °C (1013 hPa)
Vapor Pressure 1.3 hPa (81 °C)
Density 3.2 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR