2-Chloro-2-methylpropane zS, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: tert-Butyl chloride

MQ200

8.01550.0100 8.01550.1000 8.01550.9160
8.01550.0100EA 27.7 EUR
8.01550.0100 8.01550.1000 8.01550.9160
2-Chloro-2-methylpropane zS, Sigma-Aldrich®
2-Chloro-2-methylpropane
Formula: C₄H₉Cl
MW: 92.57 g/mol
Boiling Pt: 51
Melting Pt: –28 °C
Density: 0.85 g/cm³ (20 °C)
Flash Pt: –27 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000816
CAS Number: 507-20-0
EINECS: 208-066-4
UN: 1127
ADR: 3 II
Merck Index: 13,01561

Order Now

Specification Test Results

Solubility (20 °C) insoluble,(slow decomposition)
Melting Point -28 °C
Molar Mass 92.57 g/mol
Boiling Point 51 °C (1013 hPa)
Vapor Pressure 350 hPa (20 °C)
Flash Point -27 °C
Refractive Index 1.3848 (20 °C, 589 nm)
Explosion Limit 1.8 - 10.1 %(V)
Density 0.84 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 540 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR