Benzaldehyde for synthesis, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

8.01756.0100 8.01756.0500 8.01756.1000 8.01756.2500 8.01756.9028 8.01756.9025
8.01756.0100EA 42 EUR
8.01756.0100 8.01756.0500 8.01756.1000 8.01756.2500 8.01756.9028 8.01756.9025
Benzaldehyde for synthesis, Sigma-Aldrich®
Benzaldehyde
Formula: C₆H₅CHO
Boiling Pt: 179 °C (1013 hPa)
Melting Pt: –26 °C
Density: 1,049 g/cm³ (20 °C)
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00003299
CAS Number: 100-52-7
EINECS: 202-860-4
UN: 1990
ADR: 9,III
Merck Index: 13,01057
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98811dd8cad9340872b5c4b5bfb305860eb1f81a&vid=934c1b87225e77b213e66c334d8f27ef4d924847&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 3.3 g/l (20 °C)
Melting Point -26 °C
Molar Mass 106.12 g/mol
Boiling Point 179 °C (1013 hPa)
Vapor Pressure 1.3 hPa (20 °C)
Flash Point 64 °C
Refractive Index 1.545 (20 °C, 589 nm)
Explosion Limit 1.4 - 8.5 %(V)
Density 1.05 g/cm3 (20 °C)
pH 5.9 (1 g/l, H2O)
Ignition Point 190 °C
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR