2-Chlorophenol for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

8.02253.0100 8.02253.0500 8.02253.2500 8.02253.9030
8.02253.0100EA 29.8 EUR
8.02253.0100 8.02253.0500 8.02253.2500 8.02253.9030
2-Chlorophenol for synthesis, Sigma-Aldrich®
2-Chlorophenol
Formula: C₆H₅ClO
Boiling Pt: 175 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1,25 g/cm³ (20 °C)
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00002159
CAS Number: 95-57-8
EINECS: 202-433-2
UN: 2021
ADR: 6.1,III
Merck Index: 12,02173

Order Now

Specification Test Results

Solubility 28.5 g/l (20 °C)
Melting Point 7 °C
Molar Mass 128.55 g/mol
Boiling Point 174 °C (1013 hPa)
Vapor Pressure 13.3 hPa (50 °C)
Flash Point 63.9 °C
Refractive Index 1.5579 (20 °C, 589 nm)
Density 1.26 g/cm3 (20 °C)
Ignition Point 550 °C
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR