Ethyl crotonate for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

8.02668.0250 8.02668.1000
8.02668.0250EA 82.5 EUR
8.02668.0250 8.02668.1000
Ethyl crotonate for synthesis, Sigma-Aldrich®
Ethyl crotonate
Formula: C₆H₁₀O₂
Boiling Pt: 143 °C (1013 hPa)
Melting Pt: –79 °C
Density: 0,9208 g/cm³ (20 °C)
Flash Pt: 2 °C
MDL Number: MFCD00009289
CAS Number: 623-70-1
EINECS: 210-808-7
UN: 1862
ADR: 3,II
Merck Index: 13,02625

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Molar Mass 114.14 g/mol
Boiling Point 138 - 140 °C (1013 hPa)
Vapor Pressure 65 hPa (50 °C)
Flash Point 2 °C
Refractive Index 1.4248 (20 °C, 589 nm)
Density 0.92 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR