(±)-2-Chloropropionic acid for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: α-Chloropropionic Acid, (±)-2-Chloropropanoic acid, 2-Chloropropionic acid

M-Clarity™ quality level = MQ200

8.02810.0100 8.02810.0500 8.02810.9025
8.02810.0100EA 37.2 EUR
8.02810.0100 8.02810.0500 8.02810.9025
(±)-2-Chloropropionic acid for synthesis, Sigma-Aldrich®
(±)-2-Chloropropionic acid
Formula: C₃H₅ClO₂
MW: 108.52 g/mol
Boiling Pt: 180
Melting Pt: –12 °C
Density: 1.27 g/cm³ (20 °C)
MDL Number: MFCD00004224
CAS Number: 598-78-7
EINECS: 209-952-3
UN: 2511
ADR: 8 III

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -12 °C
Molar Mass 108.52 g/mol
Boiling Point 185 - 186 °C (1013 hPa)
Vapor Pressure 4 hPa (60 °C)
Flash Point 107 °C
Refractive Index 1.4345 (20 °C, 589 nm)
Density 1.27 g/cm3 (20 °C)
pH  - 1 (200 g/l, H2O, 20 °C)
Ignition Point 550 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR