Diethylamine for synthesis

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

8.03010.0100 8.03010.0500 8.03010.1000 8.03010.2500 8.03010.9050 8.03010.9700
8.03010.0100EA 23 EUR
8.03010.0100 8.03010.0500 8.03010.1000 8.03010.2500 8.03010.9050 8.03010.9700
Diethylamine for synthesis
Diethylamine
Formula: (C₂H₅)₂NH
Boiling Pt: 55,5 °C (1013 hPa)
Melting Pt: –50 °C
Density: 0,7048 g/cm³ (20 °C)
Flash Pt: –39 °C
Storage Temperature: Ambient
MDL Number: MFCD00009032
CAS Number: 109-89-7
EINECS: 203-716-3
UN: 1154
ADR: 3 (8),II
Merck Index: 13,03138
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c054fa528239af221505ef08b7c0d2503c9d1d7&vid=e1e59ef071f0c580f9eb9716f4801d314ff824c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -50 °C
Molar Mass 73.14 g/mol
Boiling Point 56 °C
Vapor Pressure 250 hPa (20 °C)
Flash Point -25 °C
Refractive Index 1.3864 (20 °C)
Explosion Limit 2.0 - 11.8 %(V)
Density 0.71 g/cm3 (20 °C)
pH 13 (100 g/l, H2O, 20 °C)
Ignition Point 310 °C
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR