Dibutyltin dichloride for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

8.04184.0250 8.04184.1000 8.04184.9030
8.04184.0250EA 95.7 EUR
8.04184.0250 8.04184.1000 8.04184.9030
Dibutyltin dichloride for synthesis, Sigma-Aldrich®
Dibutyltin dichloride
Formula: C₈H₁₈Cl₂Sn
Boiling Pt: 275 °C (1013 hPa)
Melting Pt: 37…39 °C
Density: 1,37…1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000518
CAS Number: 683-18-1
EINECS: 211-670-0
UN: 3146
ADR: 6.1,II

Order Now

Specification Test Results

Solubility 0.32 g/l (20 °C)
Melting Point 37 - 41 °C
Molar Mass 303.85 g/mol
Boiling Point 148 °C (16 hPa)
Vapor Pressure 0.0016 hPa (25 °C)
Flash Point 100 °C
Density 1.37 - 1.40 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR