N-Methyl-2-pyrrolidone (NMP), EMPLURA®, Supelco®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

8.06072.1000 8.06072.2500 8.06072.9025 8.06072.9210 8.06072.9011 8.06072.9180
8.06072.1000EA 103 EUR
8.06072.1000 8.06072.2500 8.06072.9025 8.06072.9210 8.06072.9011 8.06072.9180
N-Methyl-2-pyrrolidone (NMP), EMPLURA®, Supelco®
N-Methyl-2-pyrrolidone (NMP)

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.

Formula: C₅H₉NO
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: –24 °C
Density: 1,033 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR