1,4-Naphthoquinone for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

8.06215.0005 8.06215.0250 8.06215.1000 8.06215.9025
8.06215.0005EA 30.5 EUR
8.06215.0005 8.06215.0250 8.06215.1000 8.06215.9025
1,4-Naphthoquinone for synthesis, Sigma-Aldrich®
1,4-Naphthoquinone
Formula: C₁₀H₆O₂
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 124 °C
Density: 1,422 g/cm³ (20 °C)
Flash Pt: 141 °C
MDL Number: MFCD00001676
CAS Number: 130-15-4
EINECS: 204-977-6
UN: 2923
ADR: 8 (6.1),III
Merck Index: 12,06482

Order Now

Specification Test Results

Solubility 0.09 g/l (25 °C)
Melting Point 120 - 125 °C (sublimed)
Molar Mass 158.16 g/mol
Vapor Pressure 0.026 hPa (50 °C)
Flash Point 141 °C
Density 1.42 g/cm3 (20 °C)
pH 6.1 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR