Octylamine for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

8.06917.0250 8.06917.2500 8.06917.1000
8.06917.0250EA 53 EUR
8.06917.0250 8.06917.2500 8.06917.1000
Octylamine for synthesis, Sigma-Aldrich®
Octylamine
Formula: H₃C(CH₂)₆CH₂NH₂
Boiling Pt: 180 °C (1013 hPa)
Melting Pt: –1 °C
Density: 0,7812 g/cm³ (20 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00008247
CAS Number: 111-86-4
EINECS: 203-916-0
UN: 2734
ADR: 8 (3),I

Order Now

Specification Test Results

Solubility 0.2 g/l (25 °C)
Melting Point -1 °C
Molar Mass 129.24 g/mol
Boiling Point 178 - 179 °C (1013 hPa)
Vapor Pressure 1 hPa (20 °C)
Flash Point 60 °C
Refractive Index 1.429 (20 °C, 589 nm)
Explosion Limit 1.6 - 8.2 %(V)
Density 0.78 g/cm3 (20 °C)
Ignition Point 265 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR