Phenylhydrazine ≥97.0% for synthesis, Sigma-Aldrich®

Supplier: Merck

8.07250.0005 8.07250.0250 8.07250.1000 8.07250.0100C
8.07250.0005EA 26.5 EUR
8.07250.0005 8.07250.0250 8.07250.1000 8.07250.0100C
Phenylhydrazine ≥97.0% for synthesis, Sigma-Aldrich®
Phenylhydrazine

Phenylhydrazine for synthesis.

Formula: C₆H₈N₂
MDL Number: MFCD00007573
CAS Number: 100-63-0
Merck Index: 12,07447

Order Now

Specification Test Results

Assay (HClO₄) ≥97.0%
Density (d 20 °C/ 4 °C) 1.096 - 1.099
Identity (IR) Passes Test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt. 244 °C (1013 hPa)
Vapor Pressure 0.1 hPa (20 °C)
Flash Point 88 °C
Refractive Index 1.607 (20 °C, 589 nm)
Explosion Limit 1.1 %(V)
Density 1.10 g/cm3 (20 °C)
pH (H2O, 25 °C) alkaline,(undiluted)
Ignition Point 195 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR