Phosphorus, red stabilised for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

8.07270.0100 8.07270.0500 8.07270.2500
8.07270.0100EA 36.6 EUR
8.07270.0100 8.07270.0500 8.07270.2500
Phosphorus, red stabilised for synthesis, Sigma-Aldrich®
Phosphorus
Formula: P
MW: 30.97 g/mol
Density: ∼2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133771
CAS Number: 7723-14-0
EINECS: 231-768-7
UN: 1338
ADR: 4.1 III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 590 °C
Molar Mass 30.97 g/mol
Bulk Density 1100 kg/m3
Vapor Pressure  - 0.1 hPa (20 °C)
Density 2.34 g/cm3 (20 °C)
pH 9.5 (100 g/l, H2O, 20 °C)
Ignition Point 260 °C
Hazard Class 4.1
Hazard Class Description Flammable Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR