Triethylene glycol, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2,2'-(Ethylenedioxy)diethanol, 2,2'-(Ethane-1,2-diylbis(oxy))diethanol

M-Clarity™ quality level = MQ200

8.08245.1000 8.08245.9220
8.08245.1000EA 57.6 EUR
8.08245.1000 8.08245.9220
Triethylene glycol, Sigma-Aldrich®
Triethylene glycol
Formula: C₆H₁₄O₄
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: –7 °C
Density: 1,125 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002880
CAS Number: 112-27-6
EINECS: 203-953-2
Merck Index: 12,09802

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -7 °C
Molar Mass 150.17 g/mol
Boiling Point 285 - 295 °C (1013 hPa)
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 165 °C
Refractive Index 1.4559 (20 °C, 589 nm)
Explosion Limit 0.9 - 9.2 %(V)
Density 1.123 g/cm3 (20 °C)
pH 6.5 - 7.5 (100 g/l, H2O, 20 °C)
Ignition Point 370 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR