Trifluoroacetic acid for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

8.08260.9025 8.08260.0026 8.08260.0101 8.08260.1001 8.08260.2501 8.08260.0501
8.08260.9025EA 0 EUR
8.08260.9025 8.08260.0026 8.08260.0101 8.08260.1001 8.08260.2501 8.08260.0501
Trifluoroacetic acid for synthesis, Sigma-Aldrich®
Trifluoroacetic acid
Formula: C₂HF₃O₂
Boiling Pt: 72 °C (1013 hPa)
Melting Pt: –15 °C
Density: 1,490 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -15 °C
Molar Mass 114.02 g/mol
Boiling Point 72 °C (1013 hPa)
Vapor Pressure 141 hPa (20 °C)
Density 1.48 g/cm3 (20 °C)
pH 1 (10 g/l, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR