Triethylamine zS, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: (Diethylamino)ethane, TEA

MQ200

8.08352.0100 8.08352.0500 8.08352.1000 8.08352.2500 8.08352.9040
8.08352.0100EA 48.3 EUR
8.08352.0100 8.08352.0500 8.08352.1000 8.08352.2500 8.08352.9040
Triethylamine zS, Sigma-Aldrich®
Triethylamine
Formula: (C₂H₅)₃N
MW: 101.19 g/mol
Boiling Pt: 90
Melting Pt: –115 °C
Density: 0.729 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00009051
CAS Number: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3 (8) II
Merck Index: 13,09740
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=1c68408a0eedf1bfdb02d1b1e2337d50d449cf9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 133 g/l (20 °C)
Melting Point -115 °C
Molar Mass 101.19 g/mol
Boiling Point 90 °C (1013 hPa)
Vapor Pressure 69 hPa (20 °C)
Flash Point -11 °C
Explosion Limit 1.2 - 9.3 %(V)
Density 0.73 g/cm3 (20 °C)
Ignition Point 215 °C
Saturation concentration (20 °C) Air,no data available
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR