Squalane for synthesis, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

8.14605.0100 8.14605.1000
8.14605.0100EA 105 EUR
8.14605.0100 8.14605.1000
Squalane for synthesis, Sigma-Aldrich®
Squalane
Formula: C₃₀H₆₂
Boiling Pt: 470 °C (1013 hPa)
Melting Pt: –35 °C
Density: 0,758 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008953
CAS Number: 111-01-3
EINECS: 203-825-6
Merck Index: 12,08923

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR