p-Toluenesulphonic acid monohydrate zS, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 4-Methylbenzenesulphonic acid hydrate, Toluene-4-sulphonic acid monohydrate

MQ200

8.14725.0100 8.14725.1000 8.14725.5000 8.14725.9025
8.14725.0100EA 39.7 EUR
8.14725.0100 8.14725.1000 8.14725.5000 8.14725.9025
p-Toluenesulphonic acid monohydrate zS, Sigma-Aldrich®
p-Toluenesulphonic acid monohydrate
Formula: C₇H₈O₃S·1H₂O
MW: 190.22 g/mol
Melting Pt: 102…104 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 180
Storage Temperature: Ambient
MDL Number: MFCD00142137
CAS Number: 6192-52-5
EINECS: 203-180-0
UN: 2585
ADR: 8 III
Merck Index: 12,09671

Order Now

Specification Test Results

Solubility 670 g/l (20 °C)
Melting Point 56 °C Elimination of water of crystallisation
Molar Mass 190.22 g/mol
Bulk Density 510 kg/m3
Boiling Point 140 °C (27 hPa)
Vapor Pressure (20 °C)
Flash Point 180 °C
Density 0.5 g/cm3
pH 1 (650 g/l, H2O, 20 °C) (anhydrous substance)
Ignition Point 600 °C
Hazard Class 8
Hazard Class Description Corrosive material

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR