2-Methyl-2-butene mixture of isomers for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Trimethylethylene, Isoamylene, Isopentene

M-Clarity™ quality level = MQ200

8.14874.1000 8.14874.0025 8.14874.0250 8.14874.9025
8.14874.1000EA 101 EUR
8.14874.1000 8.14874.0025 8.14874.0250 8.14874.9025
2-Methyl-2-butene mixture of isomers for synthesis, Sigma-Aldrich®
2-Methyl-2-butene
Formula: C₅H₁₀
Boiling Pt: 39 °C (1013 hPa)
Melting Pt: –124 °C
Density: 0,6623 g/cm³ (20 °C)
Flash Pt: –45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009276
CAS Number: 513-35-9
EINECS: 247-975-0
UN: 3295
ADR: 3,I
Merck Index: 12,00649

Order Now

Specification Test Results

Solubility 0.19 g/l (25 °C)
Melting Point -134 °C
Molar Mass 70.13 g/mol
Vapor Pressure 513 hPa (20 °C)
Flash Point -45 °C
Explosion Limit 1 - 9 %(V)
Density 0.66 g/cm3 (20 °C)
Ignition Point 290 °C
Viscosity Kinematic 0.29 mm2/s (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR