L(+)-Glutamic acid for synthesis, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

8.16004.0100 8.16004.1000
8.16004.0100EA 39.7 EUR
8.16004.0100 8.16004.1000
L(+)-Glutamic acid for synthesis, Sigma-Aldrich®
L(+)-Glutamic acid
Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
Boiling Pt: 325 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1,538 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002634
CAS Number: 56-86-0
EINECS: 200-293-7
Merck Index: 13,04482
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1cb2faefcc53483f0b3bfccd6c12b611e487241&vid=a462989da97f7186f34fef7de9cad6596c2589ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 11.1 g/l (25 °C)
Melting Point 160 °C
Molar Mass 147.13 g/mol
Bulk Density 460 kg/m3
Density 1.54 g/cm3 (20 °C)
pH 3.0 - 3.5 (8.6 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR