Sodium cyanotrihydroborate for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

8.18053.0010 8.18053.0025 8.18053.0500 8.18053.0250 8.18053.0005
8.18053.0010EA 95.5 EUR
8.18053.0010 8.18053.0025 8.18053.0500 8.18053.0250 8.18053.0005
Sodium cyanotrihydroborate for synthesis, Sigma-Aldrich®
Sodium cyanotrihydroborate
Formula: CH₃BNNa
Melting Pt: 241…243 °C
Density: 1,11…1.12 g/cm³ (28 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003516
CAS Number: 25895-60-7
EINECS: 247-317-2
UN: 3134
ADR: 4.3 (6.1),I
Merck Index: 13,08680

Order Now

Specification Test Results

Solubility 2100 g/l (20 °C) (slow decomposition)
Melting Point 241 - 243 °C
Molar Mass 62.84 g/mol
Hazard Class 4.3
Hazard Class Description Dangerous When Wet Material

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR