(+)-Dimethyl-D-malate for synthesis

Supplier: Merck

8.18523.0001 8.18523.0005
8.18523.0001EA 0 EUR
8.18523.0001 8.18523.0005
(+)-Dimethyl-D-malate for synthesis
(+)-Dimethyl-D-malate
Formula: C₆H₁₀O₅
MDL Number: MFCD00066216
CAS Number: 70681-41-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR