Dipropyleneglycol monomethyl ether (mixture of isomers) for synthesis, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

8.18533.1000 8.18533.2500 8.18533.9020
8.18533.1000EA 32 EUR
8.18533.1000 8.18533.2500 8.18533.9020
Dipropyleneglycol monomethyl ether (mixture of isomers) for synthesis, Sigma-Aldrich®
Dipropyleneglycol monomethyl ether (mixture of isomers)
Formula: C₇H₁₆O₃
MDL Number: MFCD00059604
CAS Number: 34590-94-8
EINECS: 252-104-2

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 148.2 g/mol
Boiling Point 184 °C (1013 hPa)
Vapor Pressure 0.75 hPa (25 °C)
Flash Point 79 °C
Refractive Index 1.419 (25 °C, 589 nm)
Explosion Limit 1.1 - 14 %(V)
Density 0.95 g/cm3 (20 °C)
pH 6 - 7 (200 g/l, H2O, 20 °C)
Ignition Point 270 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR