Adipic acid for synthesis, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Hexanedioc acid, 1,6-Hexanedioic acid

M-Clarity™ quality level = MQ200

8.18650.0100 8.18650.1000
8.18650.0100EA 42.9 EUR
8.18650.0100 8.18650.1000
Adipic acid for synthesis, Sigma-Aldrich®
Adipic acid
Formula: HOOC(CH₂)₄COOH
Boiling Pt: 337 °C (1013 hPa)
Melting Pt: 151 °C
Density: 1,36 g/cm³ (25 °C)
Flash Pt: 196 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004420
CAS Number: 124-04-9
EINECS: 204-673-3
Merck Index: 13,00163
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58ac39e8ebd69527d91cb2d15a469a2a69506785&vid=ef907d9f9fd34903274c42b99d1d1ac1a8db6dce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 15 g/l (20 °C)
Melting Point 151 - 154 °C
Molar Mass 146.14 g/mol
Bulk Density 700 kg/m3
Boiling Point 330 °C (1013 hPa)
Vapor Pressure 0.097 hPa (18.5 °C)
Flash Point 196 °C
Density 1.360 g/cm3 (25 °C)
pH 2.7 (24.9 g/l, H2O, 25 °C)
Ignition Point 405 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR