Acetic acid 99 - 100% 99-100 zS, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

MQ200

8.18755.1000 8.18755.2500 8.18755.9025
8.18755.1000EA 46.4 EUR
8.18755.1000 8.18755.2500 8.18755.9025
Acetic acid 99 - 100% 99-100 zS, Sigma-Aldrich®
Acetic acid
Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118
Melting Pt: 17
Flash Pt: 38.5
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 (3) II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 17 °C
Molar Mass 60.05 g/mol
Boiling Point 116 - 118 °C (1013 hPa)
Vapor Pressure 15.4 hPa (20 °C)
Flash Point 44 °C
Refractive Index 1.37 (20 °C)
Explosion Limit 4 - 19.9 %(V)
Density 1.05 g/cm3 (20 °C)
pH 2.5 (50 g/l, H2O, 20 °C)
Ignition Point 485 °C
Saturation concentration 38 g/m3 (20 °C) Air
Viscosity kinematic 1.17 mm2/s (20 °C)
Evaporation number 24
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR