Ethyl palmitate for synthesis, Sigma-Aldrich®

Supplier: Merck

Synonyms: Hexadecanoic acid ethyl ester, Hexadecylic acid ethyl ester, Ethyl hexadecanoate, Palmitic acid ethyl ester

M-Clarity™ quality level = MQ200

8.18775.0005 8.18775.0250
8.18775.0005EA 27.3 EUR
8.18775.0005 8.18775.0250
Ethyl palmitate for synthesis, Sigma-Aldrich®
Ethyl palmitate
Formula: C₁₈H₃₆O₂
MW: 284.48 g/mol
Boiling Pt: 342
Melting Pt: 24
Density: 0.86 g/cm³ (20 °C)
MDL Number: MFCD00008996
CAS Number: 628-97-7
EINECS: 211-064-6

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 23 - 26 °C
Molar Mass 284.48 g/mol
Boiling Point 192 - 194 °C (13 hPa)
Flash Point 164 °C
Refractive Index 1.438 (25 °C, 589 nm)
Density 0.86 g/cm3 (20 °C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR