Tetrabutylammonium bromide ≥99.0% (by argentometric titration) for synthesis, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

8.18839.0250 8.18839.0010
8.18839.0250EA 114 EUR
8.18839.0250 8.18839.0010
Tetrabutylammonium bromide ≥99.0% (by argentometric titration) for synthesis, Sigma-Aldrich®
Tetrabutylammonium bromide
Formula: C₁₆H₃₆BrN
MW: 322.37 g/mol
Melting Pt: 100…103 °C
Density: 1.148 g/cm³ (20 °C)
MDL Number: MFCD00011633
CAS Number: 1643-19-2
EINECS: 216-699-2

Order Now

Specification Test Results

Assay (argentometric) ≥ 99.0 %
Identity (IR) passes test
Molar Mass 322.37 g/mol
Bulk Density 700 kg/m3
Density 1.15 g/cm3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR