Amantadine hydrochloride, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 1-Adamantylamine hydrochloride, 1-Aminotricyclo[3.3.1.1(3,7)]decane hydrochloride, 1-Aminotricyclo(3.3.1.1(3,7))decan HCl, 1-Aminoadamantane hydrochloride, 1-Adamantanammonium chloride, 1-Adamantanamine hydrochloride, Tricyclo[3.3.1.1(3,7)]dec-1-ylamine hydrochloride, Adamantan-1-amine hydrochloride, 1-Adamantaneammonium Chloride

M-Clarity™ quality level = MQ200

8.20015.0010 8.20015.0050
8.20015.0010EA 65.7 EUR
8.20015.0010 8.20015.0050
Amantadine hydrochloride, Sigma-Aldrich®
Amantadine hydrochloride
Formula: C₁₀H₁₈ClN
MW: 187.71 g/mol
Melting Pt: 360
Storage Temperature: Ambient
MDL Number: MFCD00074723
CAS Number: 665-66-7
EINECS: 211-560-2

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 360 °C
Molar Mass 187.71 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR