2-Ethylhexane-1,3-diol (mixture of isomers) zS, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Ethoexadiol, Octylene glycol

MQ200

8.20032.1000 8.20032.0100
8.20032.1000EA 77.4 EUR
8.20032.1000 8.20032.0100
2-Ethylhexane-1,3-diol (mixture of isomers) zS, Sigma-Aldrich®
2-Ethylhexane-1,3-diol (mixture of isomers)

Mixture of DL-and meso-form

Formula: C₈H₁₈O₂
MW: 146.23 g/mol
Boiling Pt: 244
Melting Pt: –40 °C
Density: 0.9422 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004578
CAS Number: 94-96-2
EINECS: 202-377-9

Order Now

Specification Test Results

Solubility 42 g/l (20 °C)
Melting Point -40 °C
Molar Mass 146.23 g/mol
Boiling Point 241 - 249 °C (1013 hPa)
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 136 °C
Refractive Index 1.4530 (22 °C, 589 nm)
Density 0.94 g/cm3 (20 °C)
Ignition Point 340 °C DIN 51794


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR