Trimellitic anhydride for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

8.20138.0100 8.20138.0500
8.20138.0100EA 30 EUR
8.20138.0100 8.20138.0500
Trimellitic anhydride for synthesis, Sigma-Aldrich®
Trimellitic anhydride
Formula: C₉H₄O₅
Storage Temperature: Ambient
MDL Number: MFCD00005925
CAS Number: 552-30-7
EINECS: 209-008-0

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Melting Point 164 - 168 °C
Molar Mass 192.12 g/mol
Boiling Point 240 °C (19 hPa)
Vapor Pressure 19 hPa (240 °C)
Flash Point 227 °C
Explosion Limit 1 - 7 %(V)
pH 2 (21 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR