Potassium-D-gluconate for synthesis, Sigma-Aldrich®

Supplier: Merck

Synonyms: Potassium Gluconate, Gluconic Acid Potassium Salt, D-Gluconic acid potassium salt, Potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate

M-Clarity™ quality level = MQ200

8.20601.0100 8.20601.0500
8.20601.0100EA 43.9 EUR
8.20601.0100 8.20601.0500
Potassium-D-gluconate for synthesis, Sigma-Aldrich®
Potassium D-gluconate
Formula: HOH₂C(CH(OH))₄COOK
Melting Pt: 174…176 °C
Density: 1,73 g/cm³ (20 °C)
MDL Number: MFCD00064211
CAS Number: 299-27-4
EINECS: 206-074-2
Merck Index: 12,07796

Order Now

Specification Test Results

Solubility 590 g/l (20 °C)
Melting Point 174 - 176 °C
Molar Mass 234.24 g/mol
Bulk Density 600 kg/m3
pH 7.0 - 8.3 (100 g/l, H2O, 20 °C)
Ignition Point 200 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR