Methyl hexanoate zS, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Caproic acid methyl ester, Methyl caproate, Hexanoic acid methyl ester

MQ200

8.20638.0005 8.20638.0250
8.20638.0005EA 21.7 EUR
8.20638.0005 8.20638.0250
Methyl hexanoate zS, Sigma-Aldrich®
Methyl hexanoate
Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 151
Melting Pt: –63 °C
Density: 0.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009510
CAS Number: 106-70-7
EINECS: 203-425-1
UN: 1993
ADR: 3 III

Order Now

Specification Test Results

Solubility 1.33 g/l (20 °C)
Melting Point -71 °C
Molar Mass 130.19 g/mol
Boiling Point 151 °C (1013 hPa)
Vapor Pressure 3.7 hPa (20 °C)
Flash Point 41 °C
Refractive Index 1.396 (20 °C, 589 nm)
Density 0.88 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR