4-Hexylresorcinol for synthesis, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

8.20647.0025 8.20647.0100
8.20647.0025EA 68.4 EUR
8.20647.0025 8.20647.0100
4-Hexylresorcinol for synthesis, Sigma-Aldrich®
4-Hexylresorcinol
Formula: C₁₂H₁₈O₂
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 64…67 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002284
CAS Number: 136-77-6
EINECS: 205-257-4
Merck Index: 12,04750

Order Now

Specification Test Results

Solubility 0.5 g/l (20 °C)
Melting Point 64 - 67 °C
Molar Mass 194.27 g/mol
Bulk Density 340 kg/m3
Boiling Point 333 - 335 °C (1013 hPa)
Flash Point 100 °C
pH 7.7 (0.5 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR