Iodobenzene zS, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Phenyliodide

MQ200

8.20730.0100 8.20730.0250
8.20730.0100EA 167 EUR
8.20730.0100 8.20730.0250
Iodobenzene zS, Sigma-Aldrich®
Iodobenzene
Formula: C₆H₅I
MW: 204.01 g/mol
Boiling Pt: 188
Melting Pt: –30 °C
Density: 1.838 g/cm³ (20 °C)
Flash Pt: 74
Storage Temperature: Ambient
MDL Number: MFCD00001029
CAS Number: 591-50-4
EINECS: 209-719-6
Merck Index: 13,05051

Order Now

Specification Test Results

Solubility 0.34 g/l (30 °C) (experimental)
Melting Point -30 °C
Molar Mass 204.01 g/mol
Boiling Point 187 - 189 °C
Vapor Pressure 1.41 hPa (25 °C)
Flash Point 77 °C
Refractive Index 1.6198 (20 °C, 589 nm)
Density 1.83 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR