1-Methyl-4-piperidone, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: N-Methyl-4-piperidone

MQ200

8.20828.0100
8.20828.0100EA 86.2 EUR
8.20828.0100
1-Methyl-4-piperidone, Sigma-Aldrich®
1-Methyl-4-piperidone
Formula: C₆H₁₁NO
MW: 113.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006191
CAS Number: 1445-73-4
EINECS: 215-895-5
UN: 1993
ADR: 3 III

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 113.16 g/mol
Boiling Point 60 - 62 °C (19 hPa)
Flash Point 58 °C
Refractive Index 1.4605 (20 °C, 589 nm)
Density 0.97 g/cm3 (20 °C)
pH 12 (100 g/l, H2O, 20 °C)
Synonyms N-Methyl-γ-piperidone
Hill Formula C6H11NO


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR