Sodium phenoxide trihydrate for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

8.20873.0100 8.20873.0500
8.20873.0100EA 56 EUR
8.20873.0100 8.20873.0500
Sodium phenoxide trihydrate for synthesis, Sigma-Aldrich®
Sodium phenoxide trihydrate
Formula: C₆H₅NaO·3H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149636
CAS Number: 156150-40-2
EINECS: 205-347-3
UN: 2905
ADR: 8,III

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 56 - 61 °C
Molar Mass 170.13 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR