Dodecylbenzene zS, Sigma-Aldrich®

Supplier: Merck

Synonyms: n-Dodecylbenzene, 1-Phenyldodecane

MQ200

8.20990.0025 8.20990.0100
8.20990.0025EA 162 EUR
8.20990.0025 8.20990.0100
Dodecylbenzene zS, Sigma-Aldrich®
Dodecylbenzene
Formula: C₁₈H₃₀
MW: 246.44 g/mol
Boiling Pt: 275
Melting Pt: –4 °C
Density: 0.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008974
CAS Number: 123-01-3
EINECS: 204-591-8

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 3 °C
Molar Mass 246.44 g/mol
Boiling Point 275 - 300 °C (1013 hPa)
Vapor Pressure 0.1 hPa (50 °C)
Flash Point 124 °C
Refractive Index 1.482 (20 °C, 589 nm)
Density 0.85 g/cm3 (20 °C)
Synonyms Dodecyl benzene
Hill Formula C18H30
Chemical Formula C6H5(CH2)11CH3

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR