Glycofural, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

8.21092.0100 8.21092.0250 8.21092.9010 8.21092.9025
8.21092.0100EA 41.3 EUR
8.21092.0100 8.21092.0250 8.21092.9010 8.21092.9025
Glycofural, Sigma-Aldrich®
Glycofural
Formula: C₇H₁₄O₃
MDL Number: MFCD00801146
CAS Number: 31692-85-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76a46209f55f5167d8af1ac4ba55f2bfcd86b8ed&vid=8306ea71dfa3c90670c7944d9c0fdddc7575989c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility Soluble at 20 °C
Boiling Point 176 °C (40 hPa)
Refractive Index 1.458 (27 °C, 589 nm)
Density 1,08 g/cm3 at 20 °C
Refractive index 1,458
Storage temperature +15...+25 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR