2-Thiouracil for synthesis, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-Hydroxypyrimidine-2(1H)-thione

M-Clarity™ quality level = MQ200

8.21120.0100 8.21120.0500 8.21120.1000
8.21120.0100EA 84.9 EUR
8.21120.0100 8.21120.0500 8.21120.1000
2-Thiouracil for synthesis, Sigma-Aldrich®
2-Thiouracil
Formula: C₄H₄N₂OS
Melting Pt: 340 °C
MDL Number: MFCD00006039
CAS Number: 141-90-2
EINECS: 205-508-8
Merck Index: 12,09504

Order Now

Specification Test Results

Solubility 7.9 g/l (25 °C)
Melting Point 300 °C
Molar Mass 128.14 g/mol
Bulk Density 600 kg/m3
Ignition Point 570 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR