N,N-Dimethyl-p-toluidine for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: N,N,4-Trimethylaniline, N,N-Dimethyl-4-methylaniline, 1-(Dimethylamino)-4-methylbenzol, 4,N,N-Trimethylaniline, 4-(Dimethylamino)toluene, 4-Dimethylaminotoluene

M-Clarity™ quality level = MQ200

8.22040.0005 8.22040.0250 8.22040.1000 8.22040.9025
8.22040.0005EA 22.2 EUR
8.22040.0005 8.22040.0250 8.22040.1000 8.22040.9025
N,N-Dimethyl-p-toluidine for synthesis, Sigma-Aldrich®
N,N-Dimethyl-p-toluidine
Formula: C₉H₁₃N
Boiling Pt: 211 °C (1013 hPa)
Melting Pt: –15 °C
Density: 0,9 g/cm³ (20 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00008316
CAS Number: 99-97-8
EINECS: 202-805-4
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 0.65 g/l (37 °C)
Melting Point -15 °C
Molar Mass 135.21 g/mol
Boiling Point 215 °C (1013 hPa)
Vapor Pressure 0.1 hPa (20 °C)
Flash Point 76 °C
Refractive Index 1.546 (20 °C, 589 nm)
Density 0.94 g/cm3 (20 °C)
Ignition Point 425 °C
Synonyms 4-Dimethylaminotoluene
Hill Formula C9H13N
Chemical Formula 4-(CH3)C6H4N(CH3)2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR