Sodium gluconate for synthesis, Sigma-Aldrich®

Supplier: Merck

Synonyms: D-Gluconic acid sodium salt, Sodium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconic acid, sodium salt, Sodium D-gluconate

M-Clarity™ quality level = MQ200

8.22058.0100 8.22058.1000 8.22058.2500 8.22058.9050 8.22058.9025
8.22058.0100EA 34.8 EUR
8.22058.0100 8.22058.1000 8.22058.2500 8.22058.9050 8.22058.9025
Sodium gluconate for synthesis, Sigma-Aldrich®
Sodium gluconate
Formula: HOH₂C(CH(OH))₄COONa
Melting Pt: 170…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00064210
CAS Number: 527-07-1
EINECS: 208-407-7
Merck Index: 12,08766
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eafd5b786cb23fb5c20dd64ad056ed2d80dc0186&vid=b1ffe48dafca4b8bd9b0d709ea3e986099051767&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 590 g/l (25 °C)
Melting Point 170 - 175 °C
Molar Mass 218.14 g/mol
pH 7.0 - 8.0 (100 g/l, H2O, 20 °C)
Ignition Point 200 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR