1-Hexadecanethiol for synthesis, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

8.22063.0250
8.22063.0250EA 117 EUR
8.22063.0250
1-Hexadecanethiol for synthesis, Sigma-Aldrich®
1-Hexadecanethiol
Formula: C₁₆H₃₄S
Boiling Pt: 335 °C (1013 hPa)
Melting Pt: 18…20 °C
Density: 0,8556 g/cm³ (20 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00011677
CAS Number: 2917-26-2
EINECS: 220-846-6
UN: 3334
ADR: 9,

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 18 - 20 °C
Molar Mass 258.5 g/mol
Boiling Point 185 - 190 °C (9 hPa)
Vapor Pressure  - 0.1 hPa (20 °C)
Flash Point 102 °C
Refractive Index 1.462 (20 °C, 589 nm)
Density 0.85 g/cm3 (20 °C)
Ignition Point 260 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR