1-Octadecene for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

8.22112.0100 8.22112.1000
8.22112.0100EA 51 EUR
8.22112.0100 8.22112.1000
1-Octadecene for synthesis, Sigma-Aldrich®
1-Octadecene
Formula: C₁₈H₃₆
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 17,5 °C
Density: 0,789 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009003
CAS Number: 112-88-9
EINECS: 204-012-9

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 17 - 18 °C
Molar Mass 252.48 g/mol
Boiling Point 315 °C (1013 hPa)
Vapor Pressure 1.3 hPa (20 °C)
Flash Point 154 °C
Refractive Index 1.4439 (20 °C, 589 nm)
Density 0.79 g/cm3 (20 °C)
Viscosity kinematic 3.6 mm2/s (40 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR