1,4-Dihydro-4-pyridinone for synthesis, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

8.22140.0050
8.22140.0050EA 260 EUR
8.22140.0050
1,4-Dihydro-4-pyridinone for synthesis, Sigma-Aldrich®
1,4-Dihydro-4-pyridinone
Formula: C₅H₅NO
Storage Temperature: Ambient
MDL Number: MFCD00040458
CAS Number: 108-96-3
EINECS: 203-633-2

Order Now

Specification Test Results

Solubility Soluble (20 °C)
Melting Point 140 - 147 °C
Molar Mass 95.1 g/mol
Bulk Density 600 kg/m3
Boiling Point 170 - 172 °C (0.03 hPa)
Flash Point 212 °C
pH 7 (50 g/l, H2O, 20 °C)
Ignition Point 515 °C
Bulk density 600 kg/m3
Assay (HClO4) ≥95%
Sulfated ash ≤5%
Identity (IR) Passes test
Storage temperature +15...+25 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR